Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Venezuelan Eq. Encephalitis Mouse anti-Human, Clone: 1A3B.7, MilliporeSigma™

Catalog No. MAB8755MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8755MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8755MI Supplier MilliporeSigma Supplier No. MAB8755
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Venezuelan Eq. Encephalitis using this Anti-Venezuelan Eq. Encephalitis Antibody, clone 1A3B-7 validated for use in ELISA, immunofluoresence, immunohistochemistry, NEUT.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Venezuelan Eq. Encephalitis
Applications ELISA, Immunofluorescence, Immunohistochemistry, Neutralization
Classification Monoclonal
Clone 1A3B.7
Conjugate Unconjugated
Formulation Purified
Host Species Mouse
Immunogen VEE
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.