Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Versican Rabbit anti-Mouse, Polyclonal, MilliporeSigma™

Catalog No. AB1033MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB1033MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB1033MI Supplier MilliporeSigma Supplier No. AB1033
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Versican Antibody, a.a. 1360-1439 of mouse versican is an antibody against Versican for use in immunoprecipitation, western blot, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Versican
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation The immunoglobulin was affinity purified against the recombinant antigen and is presented in PBS. Anti-GST reactivity has not been adsorbed. Liquid in PBS.
Gene Symbols ACAN; MSK16; SEDK; CSPCP; AGCAN; AGC1; CSPG1; CSPGCP
Host Species Rabbit
Immunogen GST fusion protein containing amino acids 1360-1439 of mouse versican.
Purification Method Immunoaffinity purified
Quantity 50 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_001135.2; NM_013227.2
Target Species Mouse
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.