Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vesicular Glutamate Transporter 1 Guinea Pig anti-Rat, Polyclonal, MilliporeSigma™

Catalog No. AB5905MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB5905MI 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB5905MI Supplier MilliporeSigma Supplier No. AB5905
Add to Cart
Add to Cart

Guinea Pig Polyclonal Antibody

This Anti-Vesicular Glutamate Transporter 1 Antibody is validated for use in Immunohistochemistry for the detection of Vesicular Glutamate Transporter 1 with more than 55 product citations.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vesicular Glutamate Transporter 1
Applications Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Formulation Guinea pig Serum in 0.1% Sodium Azide.
Gene Symbols VGluT1; Solute carrier family 17 member 7; Brain-specific Na(+)-dependent inorganic phosphate cotransporter; Slc17a7; Bnpi
Host Species Guinea Pig
Immunogen Synthetic peptide from rat VGLUT1 protein with no overlap to VGLUT2. The immunogen is available as AG208.
Purification Method Unpurified
Quantity 50 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_446311
Target Species Rat
Content And Storage −20°C in undiluted aliquotes for 6 months. Avoid repeated freeze/thaw cycles.
Form Serum
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.