Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vesicular Glutamate Transporter 2 Mouse anti-Mouse, Rat, Human, AlexaFluor™488, Clone: 8G9.2, MilliporeSigma™

Catalog No. MAB5504A4MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5504A4MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5504A4MI Supplier MilliporeSigma Supplier No. MAB5504A4
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Vesicular Glutamate Transporter 2 Antibody, Alexa Fluor 488 is validated for use in immunohistochemistry for the detection of Vesicular Glutamate Transporter 2.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vesicular Glutamate Transporter 2
Applications Immunohistochemistry
Classification Monoclonal
Clone 8G9.2
Conjugate Alexa Fluor 488
Formulation Purified mouse monoclonal IgG1 conjugated to Alexa Fluor™ 488 in PBS with 0.1% sodium azide and 15mg/mL BSA.
Gene Accession No. NP_445879
Gene Symbols Slc17a6, Dnpi, Vglut2, VGLUT2
Host Species Mouse
Immunogen Recombinant protein from rat VGLUT2.
Purification Method Protein A purified
Quantity 100 μL
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C in undiluted aliquots for up to 6 months. Protect from Light.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.