Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vesicular Glutamate Transporter 2 Mouse anti-Human, Mouse, Rat, Clone: 8G9.2, MilliporeSigma™

Catalog No. MAB5504MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB5504MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB5504MI Supplier MilliporeSigma Supplier No. MAB5504
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Vesicular Glutamate Transporter 2 using this Anti-Vesicular Glutamate Transporter 2 Antibody validated for use in western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vesicular Glutamate Transporter 2
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 8G9.2
Conjugate Unconjugated
Formulation Mouse monoclonal in PBS containing 0.1% sodium azide.
Gene Accession No. NM_020346.1
Gene Symbols SLC17A6, DNPI, VGLUT2
Host Species Mouse
Immunogen Recombinant protein from rat VGLUT2.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Stable for 1 year at from date of receipt.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.