Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vesicular Monoamine Transporter 1 Rabbit anti-Mouse, Rat, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
Supplier: MilliporeSigma AB1597P
Description
Anti-Vesicular Monoamine Transporter 1 Antibody detects level of Vesicular Monoamine Transporter 1 and has been published and validated for use in ELISA, immunohistochemistry and western blotting.
Specifications
| Vesicular Monoamine Transporter 1 | |
| Polyclonal | |
| Affinity purified immunoglobulin. Liquid in PBS with 0.1% BSA. | |
| Rabbit | |
| Affinity Purified | |
| Primary | |
| Mouse, Rat |
| ELISA, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| SLC18A1; VAT1; CGAT; VMAT1 | |
| A 12 amino acid C-terminal peptide sequence from rat VMAT1 (Liu et al. 1992). This peptide is predicted to be cytoplasmic. | |
| 50 μg | |
| NM_003053.1 | |
| −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction