Learn More
Vesicular Monoamine Transporter 2 Rabbit anti-Rat, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Vesicular Monoamine Transporter 2 |
| Applications | ELISA, Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Liquid in PBS with 0.1% BSA. |
| Gene Symbols | SLC18A2; SVAT; MGC26538; SVMT; VAT2; MGC120477; VMAT2; MGC120478 |
| Host Species | Rabbit |
| Immunogen | A 13 amino acid C-terminal peptide sequence from rat VMAT2 (Liu et al. 1992). This peptide is predicted to be cytoplasmic. |
| Purification Method | Affinity Purified |
| Quantity | 50 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.