Learn More
MilliporeSigma™ anti-VGLUT2, Polyclonal, (Chemicon)
Supplier: MilliporeSigma™ AB2251/DEL
Description
VGLUT2 is a sodium dependent inorganic phosphate cotransporter that is involved in the calcium dependent glutamate release from astrocytes. It is expressed in only a subset of neurons, localized to synaptic vesicles, at synapses exhibiting classical excitatory features. VGLUT2 mRNA is found in brain regions that lack VGLUT1, a related protein also displaying sodium dependent inorganic phosphate cotransporter activity.
Immunohistochemistry, Western Blotting
Specifications
VGLUT2 | |
Unconjugated | |
Recombinant GST-tagged. | |
RUO | |
Primary | |
Serum |
Polyclonal | |
Guinea Pig | |
50 μL | |
Neuroscience | |
Mouse, Rat |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.