Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VHL Mouse anti-Human, Alexa Fluor 647, Clone: S2-647, BD

Catalog No. BDB564184 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB564184 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB564184 Supplier BD Biosciences Supplier No. 564184
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The S2-647 monoclonal antibody recognizes the Von Hippel-Lindau (VHL) protein. The VHL protein is expressed within the cytosol and nuclei of cells from a variety of tissues. It serves as a tumor suppressor and transcriptional repressor that is highly conserved from Drosophila to mammalian species. VHL binds to Elongin B and C subunits and other proteins as part of a E3 ubiquitin ligase complex that targets a variety of proteins, including HIF-1 alpha and Beta-2 adrenergic receptors, for proteasomal degradation. Aberrant VHL expression is associated with a number of diseases including Von Hippel-Lindau disease, a familial cancer syndrome, that predisposes individuals to hemangioblastomas of the central nervous system and retina, pheochromocytoma and renal cell carcinoma. Three distinct VHL isoforms ranging in molecular weight from approximately 19-30 kDa have been described.

TRUSTED_SUSTAINABILITY

Specifications

Antigen VHL
Applications Flow Cytometry
Classification Monoclonal
Clone S2-647
Conjugate Alexa Fluor 647
Description VHL1; pVHL; Elongin binding protein; Protein G7; HRCA1; RCA1
Host Species Mouse
Immunogen Human VHL Recombinant Protein
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG2b κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.