Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VHLL, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89206543 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-206-543 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-206-543 Supplier Abnova Corporation Supplier No. H00391104K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human VHLL peptide using ARM Technology.

Von Hippel-Lindau (VHL) tumor suppressor protein is a component of an E3 ubiquitin ligase complex that the selectively ubiquitinates the alpha subunit of the hypoxia-inducible factor (HIF) transcription factor for proteasome-mediated degradation. Inactivation of VHL causes VHL disease and sporadic kidney cancer. This gene encodes a VHL homolog that lacks one of two key domains necessary for VHL function. It binds HIF alpha but fails to recruit the E3 ubiquitin ligase complex, and therefore functions as a dominant-negative VHL and a protector of HIF alpha. This gene is intronless and predominantly expressed in the placenta, and may contribute to the regulation of oxygen homeostasis and neovascularization during placenta development. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen VHLL
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description von Hippel-Lindau tumor suppressor-like
Formulation In 1X PBS, pH 7.4
Gene VHLL
Gene Accession No. VHLL
Gene Alias VLP
Gene Symbols VHLL
Host Species Rabbit
Immunogen A synthetic peptide of human VHLL
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 391104
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.