Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Impacto™ Anti-Vibration Cowhide Leather Air Glove™

Catalog No. p-7084598
Click to view available options
Size:
Small
Medium
Large
X-Large
This item is not returnable. View return policy
This item is not returnable. View return policy

Full finger leather air gloves utilize patented technology in the palm, fingers, and thumb to provide maximum vibration and impact protection.

  • Full finger, all leather style for maximum durability and protection
  • Durable grain cowhide leather and air padding construction
  • Helps prevent hand-arm vibration syndrome (HAVS)
  • For use with heavy duty tools such as jackhammers and grinding
  • Meets anti-vibration glove standards EN ISO 10819 and ANSI S2.73-2002 (R 2007), ISO 10819:1996

Specifications

Certifications/Compliance ANSI S2.73-2002 (R 2007)/ISO 10819
Color Gold
Hand Specific
Latex Free Yes
Material Cowhide Leather
Product Type Anti-vibration Gloves
Size Small
Glove Cuff Style Open
Hem Color Brown
Series BG650
Thumb Style Wing
Length (English) 12 in.
Length (Metric) 30.5 cm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.