Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

villin 1, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89018354 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-018-354 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-018-354 Supplier Abnova Corporation Supplier No. H00007429D01P
Add to Cart
Add to Cart

Rabbit polyclonal antibody raised against a full-length human VIL1 protein.

This gene encodes a member of a family of calcium-regulated actin-binding proteins. This protein represents a dominant part of the brush border cytoskeleton which functions in the capping, severing, and bundling of actin filaments. Two mRNAs of 2.7kb and 3.5kb have been observed; they result from utilization of alternate poly-adenylation signals present in the terminal exon. [provided by RefSeq

Sequence: MTKLSAQVKGSLNITTPGLQIWRIEAMQMVPVPSSTFGSFFDGDCYIILAIHKTASSLSYDIHYWIGQDSSLDEQGAAAIYTTQMDDFLKGRAVQHREVQGNESEAFRGYFKQGLVIRKGGVASGMKHVETNSYDVQRLLHVKGKRNVVAGEVEMSWKSFNRGDVFLLDLGKLIIQWNGPESTRMERLRGMTLAKEIRDQERGGRTYVGVVDGENELASPKLMEVMNHVLGKRRELKAAVPDTVVEPALKAALKLYHVSDSEGNLVVREVATRPLTQDLLSHEDCYILDQGGLKIYVWKGKKANEQEKKGAMSHALNFIKAKQYPPSTQVEVQNDGAESAVFQQLFQKWTASNRTSGLGKTHTVGSVGEGQAGAVREPGSRSWARRATWSTTHPPSLTCIFNEDFYAGSGLVLADGDVDKL

Specifications

Antigen villin 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against a full-length human VIL1 protein.
Formulation PBS with no preservative; pH 7.4
Gene VIL1
Gene Accession No. BC017303.2
Gene Alias D2S1471/VIL
Gene Symbols VIL1
Host Species Rabbit
Immunogen VIL1 (AAH17303.1, 1 a.a. ∼ 421 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7429
Target Species Human, Mouse
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.