Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Villin Mouse anti-Human, Unlabeled, Clone: 12, BD

Mouse Monoclonal Antibody

Supplier:  BD Biosciences 610358

Catalog No. BDB610358


Only null left
Add to Cart

Description

Description

The isolated intestinal microvillus cytoskeleton (core) consists of four major proteins: actin, villin, fimbrin, and brush border myosin-I. These proteins can assemble in vitro into structures resembling native microvillus cores. Of these components, villin, and brush border myosin-I show tissue-specific expression, so they may be involved in the morphogenesis of intestinal microvilli. Found in association with the microvillar actin bundles of the intestinal brush border, villin is a 95kDa protein composed of two very similar domains of approximately 44kDa each, the core, and a C-terminal domain of 8kDa, the headpiece. The core has been shown to contain villin's Ca2+ regulated capping, nucleating, and severing activities, but it cannot induce the formation of microfilament bundles without the headpiece. Villin is a useful differentiation marker of early embryogenesis and may be useful in diagnosis and follow-up of colorectal cancers. It has been demonstrated that villin is necessary for both the cytoskeletal and membrane protein organization of a functional brush border.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Villin
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Human
IgG1
Immunofluorescence, Immunohistochemistry, Western Blot
12
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only.