Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Goat anti-Human, Mammals, Polyclonal, MilliporeSigma™
Goat Polyclonal Antibody
Supplier: MilliporeSigma AB1620
Description
Anti-Vimentin Antibody detects level of Vimentin and has been published and validated for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
Specifications
Vimentin | |
Polyclonal | |
Neat goat Serum. Liquid containing 0.1% sodium azide. | |
Goat | |
Unpurified | |
Primary | |
Human, Mammalia | |
Serum |
Immunocytochemistry, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
VIM; FLJ36605; Vimentin.; vimentin | |
Vimentin purified from cultured human foreskin fibroblasts. | |
500 mL | |
NM_003380.2 | |
−20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction