Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Mouse anti-Human, Chicken, Rat, Lizard, Clone: 3CB2, MilliporeSigma™

Catalog No. MABT121MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABT121MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABT121MI Supplier MilliporeSigma Supplier No. MABT121
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Vimentin Antibody, clone 3CB2 is an antibody against Vimentin for use in western blot, immunocytochemistry, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vimentin
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3CB2
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.
Gene Accession No. NP_001041541
Gene Symbols VIM
Host Species Mouse
Immunogen Vimentin extracted from chicken embryonic brain tissue.
Purification Method Ion Exchange
Quantity 100 μL
Primary or Secondary Primary
Target Species Chicken, Human, Lizard, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgM κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.