Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Vimentin Mouse anti-Human, Mouse, Clone: LN-6, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MAB1681
Description
Anti-Vimentin Antibody, clone LN-6 detects level of Vimentin and has been published and validated for use in immunohistochemistry (paraffin).
Specifications
Vimentin | |
Monoclonal | |
Unconjugated | |
NM_003380.2 | |
Mouse | |
1 mL | |
Human, Mouse | |
Supernatant |
Immunohistochemistry (Paraffin) | |
LN-6 | |
Liquid tissue culture supernatant. | |
VIM, FLJ36605, Vimentin., vimentin | |
Nuclei from adult human thymic biopsy specimens. | |
Primary | |
−20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. | |
IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction