Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Mouse, Unlabeled, Clone: RV202, BD

Catalog No. BDB550513 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB550513 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB550513 Supplier BD Biosciences Supplier No. 550513
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Intermediate filaments (IF) are a subset of cytoskeletal proteins which function to give overall structural integrity to the plasma membrane as well to organize cells into specific tissues. IF proteins can be divided into six major types based upon the similarity in sequence. Vimentin belongs to the type III category of IF proteins which are expressed in leukocytes, blood vessel endothelial cells, some epithelial cells, and mesenchymal cells. Vimentin is also expressed together with several other IF proteins during the early stages of development. Vimentin is
exchanged for the tissue-specific intermediate filament type as differentiation proceeds. Vimentin migrates in SDS/PAGE as a ∽57kDa protein.

Host Species: Mouse
Clone: RV202
Isotype: IgG1
Species Reactivity: Human
Immunogen: Purified Cow Lens Vimentin

Electron Microscopy, Immunofluorescence, Immunohistochemistry, Western Blotting

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vimentin
Applications Western Blot
Classification Monoclonal
Clone RV202
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified Cow Lens Vimentin
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Bovine, Canine, Chicken, Goat, Hamster, Human, Monkey, Mouse, Rabbit, Rat
Content And Storage Store undiluted at 4°C
Isotype IgG1
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.