Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Mouse anti-Bovine, Canine, Chicken, Feline, Human, Monkey, Pig, Rat, Clone: V9, MilliporeSigma™

Catalog No. MAB3400MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
40 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3400MI 40 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3400MI Supplier MilliporeSigma Supplier No. MAB3400
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Vimentin Antibody, clone V9 is an antibody against Vimentin for use in immunocytochemistry, immunohistochemistry, immunohistochemistry (paraffin) and western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vimentin
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone V9
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1 presented in 0.02MPhosphate buffer, 0.25MNaCl with 0.1% sodium azide.
Gene Accession No. NM_003380.2
Gene Symbols VIM, FLJ36605
Host Species Mouse
Immunogen Purified vimentin from pig eye lens
Purification Method Protein A purfied
Quantity 40 mg
Primary or Secondary Primary
Target Species Bovine, Canine, Chicken, Feline, Human, Monkey, Pig, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.