Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Mouse anti-Amphibian, Bovine, Canine, Chicken, Human, Monkey, Clone: VIM 3B4, MilliporeSigma™

Catalog No. CBL202MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CBL202MI 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CBL202MI Supplier MilliporeSigma Supplier No. CBL202
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Vimentin Antibody, clone VIM 3B4 is validated for use in ELISA, immunofluoresence, immunohistochemistry (paraffin), western blotting for the detection of Vimentin.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vimentin
Applications ELISA, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone VIM 3B4
Conjugate Unconjugated
Formulation Protein A Purified immunoglobulin presented as lyophilized material. Reconstitute with 1ml distilled water. Final solution is PBS, pH 7.4, containing 0.5% BSA and 0.09% sodium azide.
Gene Accession No. NM_003380.2
Gene Symbols VIM, FLJ36605, Vimentin., vimentin
Host Species Mouse
Immunogen Vimentin purified from bovine lens
Purification Method Protein A purfied
Quantity 50 μg
Primary or Secondary Primary
Target Species Amphibian, Bovine, Canine, Chicken, Human, Monkey
Content And Storage 2-8°C for up to 12 months.
Form Purified
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.