Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Vimentin/LN6 Clone: V-9,

Catalog No. IF01100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
IF01100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. IF01100UG Supplier MilliporeSigma™ Supplier No. IF01100UG

Mouse Monoclonal Antibody

Specifically detects Vimentin/LN6 Clone: V-9 in Chicken, Human, Porcine, Rat samples, and it is validated for Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immuno Blotting

Intermediate (10 nm) filaments are present in, and provide information concerning the origin of most vertebrate cells. At present, five major cell types can be distinguished by their intermediate filament components. These include epithelial cells (cytokeratins), neurons (neurofilaments), glial cells (GFA), skeletal, visceral and certain vascular smooth muscle cells (desmin) and various nonepithelial cells, including cells of mesenchymal origin (vimentin). A few well-defined cell types coexpress vimentin along with one other intermediate filament type. Vimentin (Ab-1) stains the vimentin subclass of intermediate filaments in human tissues.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vimentin/LN6
Applications Immunoblot, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone V-9
Conjugate Unconjugated
Formulation In 50mM sodium phosphate buffer, 0.2% gelatin.
Host Species Mouse
Immunogen purified porcine vimentin
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽58kDa vimentin protein. Does not cross-react with closely related intermediate filament proteins including desmin and GFA.
Target Species Chicken, Human, Pig, Rat
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.