Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vimentin Mouse anti-Human, PE, Clone: RV202, BD

Catalog No. BDB562337 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB562337 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB562337 Supplier BD Biosciences Supplier No. 562337
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Intermediate filaments (IF) are a subset of cytoskeletal proteins which function to give overall structural integrity to the plasma membrane as well to organize cells into specific tissues. IF proteins can be divided into six major types based upon the similarity in sequence. Vimentin belongs to the type III category of IF proteins which are expressed in leukocytes, blood vessel endothelial cells, some epithelial cells, and mesenchymal cells. Vimentin is also expressed together with several other IF proteins during the early stages of development. Vimentin is exchanged for the tissue-specific intermediate filament type as differentiation proceeds. Vimentin migrates in SDS/PAGE as a ∽57 kDa protein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen Vimentin
Applications Flow Cytometry
Classification Monoclonal
Clone RV202
Conjugate PE
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Purified Cow Lens Vimentin
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.