Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ anti-Vinculin Clone: V284 or VLN01,

Catalog No. CP74100UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
Conjugate:
Unconjugated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Conjugate
CP74100UG 100 μg Unconjugated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CP74100UG Supplier MilliporeSigma™ Supplier No. CP74100UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Specifically detects Vinculin Clone: V284 or VLN01 in Human samples, and it is validated for Immunohistochemistry (Paraffin), Immunoprecipitation

Vinculin is an attachment protein involved in the indirect binding of intracellular actin filaments to extracellular fibronectin. Vinculin binds to actin capping protein and α-actinin, which in turn binds to actin. Also, vinculin binds to talin, which in turn binds to the intracellular domain of the fibronectin receptor. Vinculin has been associated with tumor suppressor activity. Vinculin is widely distributed in tissues and expressed where smooth muscle actin and fibroblasts attach to the extracellular matrix. Vinculin has been found in all adherens junctions, while the larger splice variant, meta-vinculin, is found only in smooth and cardiac muscle.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vinculin
Applications Immunohistochemistry (Paraffin), Immunoprecipitation
Classification Monoclonal
Clone V284 or VLN01
Conjugate Unconjugated
Formulation In 10mM PBS, 0.2% BSA, pH 7.4.
Host Species Mouse
Immunogen semi-purified, human vinculin
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Recognizes the ∽130 (vinculin) and ∽150kDa (meta-vinculin) proteins in skin tissue.
Target Species Human
Content And Storage 2°C to 8°C
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.