Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vinculin Mouse anti-Bovine, Human, Mouse, Monkey, Pig, Rabbit, Clone: VIIF9 (7F9), MilliporeSigma™

Catalog No. MAB3574MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB3574MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB3574MI Supplier MilliporeSigma Supplier No. MAB3574
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Vinculin Antibody, clone VIIF9 (7F9) is an antibody against Vinculin for use in immunoprecipitation, western blot, immunocytochemistry, immunohistochemistry (paraffin).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vinculin
Applications Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone VIIF9 (7F9)
Conjugate Unconjugated
Formulation Liquid in 0.02M Phosphate buffer with 0.25M NaCl and 0.1% sodium azide.
Gene Accession No. NM_003373.3; NM_014000.2
Gene Symbols VCL, MVCL, Vinculin, Metavinculin, OTTHUMP00000043885, CMD1W, vinculin
Host Species Mouse
Immunogen Crude smooth muscle extract from normal human adult uterus.
Quantity 100 μg
Primary or Secondary Primary
Target Species Bovine, Human, Monkey, Mouse, Pig, Rabbit
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.