Learn More
Vitamin K dependent protein S Rabbit anti-Mouse, Human, Horse, Chimpanzee, Monkey, Rat, Rhesus Macaque, Polyclonal, MilliporeSigma™
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Antigen | Vitamin K dependent protein S |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Purified rabbit polyclonal containing 0.1M Tris-glycine, pH 7.4, 0.150mM NaCl and 0.05% sodium azide. |
| Gene Symbols | PROS; PS21; PS22; PS23; PS24; PS25; PSA |
| Host Species | Rabbit |
| Immunogen | This antibody is generated from rabbits immunized with a KLH-conjugated linear peptide corresponding to residues surrounding the mature peptide region of Vitamin K dependent protein S. |
| Purification Method | Affinity Purified |
| Quantity | 100 μg |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.