Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vitronectin Mouse anti-Human, Clone: BV2, MilliporeSigma™

Catalog No. MAB1945MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1945MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB1945MI Supplier MilliporeSigma Supplier No. MAB1945
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Vitronectin Antibody, clone BV2 is an antibody against Vitronectin for use in ELISA, flow cytometry, immunoprecipitation, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vitronectin
Applications ELISA, Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone BV2
Conjugate Unconjugated
Formulation Purified immunoglobulin by Protein A chromatography. Liquid in PBS containing 0.02% sodium azide and 0.1% BSA.
Gene Accession No. NM_000638.3
Gene Symbols VTN, S-protein, epibolin, VNT, VN, V75
Host Species Mouse
Immunogen Purified human plasma vitronectin (Preissner, 1985).
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 12 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.