Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VLDL-R Mouse anti-Mouse, Human, Rat, Clone: 6A6, MilliporeSigma™

Catalog No. MABS25MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABS25MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABS25MI Supplier MilliporeSigma Supplier No. MABS25

Mouse Monoclonal Antibody

Anti-VLDL-R Antibody, clone 6A6 is an antibody against VLDL-R for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VLDL-R
Applications Western Blot
Classification Monoclonal
Clone 6A6
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1λ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001018066
Gene Symbols VLDLRCH, VLDL-R, CARMQ1, CHRMQ1, VLDLR
Host Species Mouse
Immunogen KLH-conjugated linear peptide corresponding to the C-terminus of human VLDL-R.
Purification Method Protein G
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 λ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.