Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

vanin 3, Mouse, Polyclonal Antibody, Abnova™

Catalog No. 89004664 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-004-664 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-004-664 Supplier Abnova Corporation Supplier No. H00055350A01
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a partial recombinant VNN3.

This gene is the central gene in a cluster of three vanin genes on chromosome 6q23-q24. The open reading frame is disrupted by a frameshift, and all splice variants that have been described are candidates for nonsense-mediated decay (NMD). Consequently, it is unlikely that this gene expresses a protein in vivo, so it is classified as a pseudogene. Extensive alternative splicing has been described; the two most common variants are represented as RefSeqs. [provided by RefSeq

Sequence: ARYHKYNLFAPEIQFDFPKDSELVTFDTPFGKFGIFTCFDIFSHDPAVVVVDEFQLTAFSTPQHGTTRCPSSRLFPSIQHGPRPWESIYLLQIPTTPACT

Specifications

Antigen vanin 3
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a partial recombinant VNN3.
Formulation 50% glycerol
Gene VNN3
Gene Accession No. NM_018399
Gene Alias HSA238982/MGC171203/PAGEL-beta/PAGEL-eta/PAGEL-zeta
Gene Symbols VNN3
Host Species Mouse
Immunogen VNN3 (NP_060869.2, 175 a.a. ∼ 274 a.a) partial recombinant protein with GST tag.
Quantity 50 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 55350
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Serum
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.