Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

von Willebrand Factor Rabbit anti-Human, Mouse, Rat, Polyclonal, MilliporeSigma™

Catalog No. AB7356MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AB7356MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AB7356MI Supplier MilliporeSigma Supplier No. AB7356
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-von Willebrand Factor, Cat. No. AB7356, is a rabbit polyclonal antibody that detects von Willebrand Factor and has been tested for use in ELISA, Immunohistochemistry, and Western Blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen von Willibrand Factor
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit immunoglobulin in buffer containing 0.02M PBS pH 7.6, 0.25M NaCl with 0.1% sodium azide.
Gene Symbols VWF; F8VWF; F8; vWF; VWD
Host Species Rabbit
Immunogen Human von Willebrand Factor purified from plasma.
Purification Method Protein A purfied
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NM_000552.3
Target Species Human, Mouse, Rat
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.