Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

visinin-like 1, Mouse, Clone: 2F1-E3, Abnova™

Catalog No. 89014212 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-014-212 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-014-212 Supplier Abnova Corporation Supplier No. H00007447M01
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant VSNL1.

This gene is a member of the visinin/recoverin subfamily of neuronal calcium sensor proteins. The encoded protein is strongly expressed in granule cells of the cerebellum where it associates with membranes in a calcium-dependent manner and modulates intracellular signaling pathways of the central nervous system by directly or indirectly regulating the activity of adenylyl cyclase. Alternatively spliced transcript variants have been observed, but their full-length nature has not been determined. [provided by RefSeq

Sequence: MGKQNSKLAPEVMEDLVKSTEFNEHELKQWYKGFLKDCPSGRLNLEEFQQLYVKFFPYGDASKFAQHAFRTFDKNGDGTIDFREFICALSITSRGSFEQKLNWAFNMYDLDGDGKITRVEMLEIIEAIYKMVGTVIMMKMNEDGLTPEQRVDKIFSKMDKNKDDQITLDEFKEAAKSDPSIVLLLQCDIQK

Specifications

Antigen visinin-like 1
Applications ELISA, Western Blot
Classification Monoclonal
Clone 2F1-E3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant VSNL1.
Formulation PBS with no preservative; pH 7.4
Gene VSNL1
Gene Accession No. BC022012
Gene Alias HLP3/HPCAL3/HUVISL1/VILIP/VILIP-1
Gene Symbols VSNL1
Host Species Mouse
Immunogen VSNL1 (AAH22012, 1 a.a. ∼ 191 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Stem Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 7447
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.