Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vsp4/SKD1 Mouse anti-Human, Mouse, Clone: 5E6.1, MilliporeSigma™

Catalog No. MABC83MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABC83MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABC83MI Supplier MilliporeSigma Supplier No. MABC83
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-Vsp4/SKD1 Antibody, clone 5E6.1 is an antibody against Vsp4/SKD1 for use in Western Blotting, immunocytochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Vsp4/SKD1
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 5E6.1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG1κ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_004860
Gene Symbols VPS4B, SKD1, VPS42, MIG1
Host Species Mouse
Immunogen Linear peptide corresponding to human Vps4/SKD1.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.