Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ VSV-G Tag Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA130138
Description
PA1-30138 detects VSV-G from rabbit samples.
Vesicular stomatitis virus (VSV), an enveloped RNA virus from the Rhabdoviridae family, is released from the plasma membrane of host cells by a process called budding. The glycoprotein (VSV G) contains a domain in its extracellular membrane-proximal stem that appears to be needed for efficient VSV budding: Sequence: YTDIEMNRLGK.
Specifications
VSV-G Tag | |
Polyclonal | |
Unconjugated | |
vesicular stomatitis virus; VSIVgp3; VSV epitope tag; VSV tag; vsvg; VSVG epitope tag; VSV-G epitope tag; VSVG tag; YTDIEMNRLGK | |
Rabbits were immunized with YTDIEMNRLGK (VSV-g epitope) conjugated with KLH. | |
100 μg | |
Primary | |
4°C | |
Liquid | |
Tag |
ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 0.1% sodium azide; pH 7.2 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
Tag | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction