Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

vWF Mouse anti-Human, Unlabeled, Clone: 2F2-A9, BD

Catalog No. BDB555849 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB555849 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB555849 Supplier BD Biosciences Supplier No. 555849
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reacts with von Willebrand Factor (vWF), a multimeric plasma glycoprotein synthesized by endothelial cells and megakaryocytes. In plasma, vWF circulates as multimers which range from dimers of about 500kDa to multimers of 20000kDa. It functions as a carrier for Factor VIII and is a mediator of initial platelet adhesion to subendothelium and platelet-platelet interaction. Platelets bind to vWF through the platelet GPIb/IX (CD42b/CD42a) and GPIIb/IIIa (CD41/CD61) complexes This antibody is effective in detecting vWF by ELISA, with no evidence of cross reactivity with fibronectin or fibrinogen.

Host Species: Mouse
Clone: 2F2-A9
Isotype: IgG1 κ
Species Reactivity: Human

Immunohistochemistry (Frozen)

TRUSTED_SUSTAINABILITY

Specifications

Antigen vWF
Applications Immunohistochemistry (Frozen)
Classification Monoclonal
Clone 2F2-A9
Concentration 0.5mg/mL
Conjugate Unconjugated
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Host Species Mouse
Purification Method Affinity Purified
Quantity 0.5 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store undiluted at 4°C
Isotype IgG1 κ
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.