Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

vWF (von Willebrand Factor) Mouse anti-Human, Clone: GMA-022, MilliporeSigma™

Catalog No. 05861MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-861-MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-861-MI Supplier MilliporeSigma Supplier No. 05861
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-vWF (von Willebrand Factor) Antibody, clone GMA-022 is an antibody against vWF (von Willebrand Factor) for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen vWF (von Willebrand Factor)
Applications Western Blot
Classification Monoclonal
Clone GMA-022
Conjugate Unconjugated
Gene Accession No. NM_000552
Gene Symbols VWF, F8, vWF, VWD, F8VWF
Host Species Mouse
Immunogen Purified human von Willebrand Factor
Purification Method DEAE purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.