Learn More
WASF3, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human WASF3 peptide using ARM Technology.
Supplier: Abnova Corporation H00010810K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
This gene encodes a member of the Wiskott-Aldrich syndrome protein family. The gene product is a protein that forms a multiprotein complex that links receptor kinases and actin. Binding to actin occurs through a C-terminal verprolin homology domain in all family members. The multiprotein complex serves to tranduce signals that involve changes in cell shape, motility or function. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
WASF3 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
WASF3 | |
WASF3 | |
A synthetic peptide of human WASF3 | |
RUO | |
10810 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
WAS protein family, member 3 | |
WASF3 | |
Brush-1, KIAA0900, SCAR3, WAVE3 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.