Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WASH complex subunit 7 (SWIP) Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. ABT70MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABT70MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ABT70MI Supplier MilliporeSigma Supplier No. ABT70
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect WASH complex subunit 7 (SWIP) using this Anti-WASH complex subunit 7 (SWIP) Antibody validated for use in immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WASH complex subunit 7 (SWIP)
Applications Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Symbols KIAA10331
Host Species Rabbit
Immunogen GST-tagged recombinant protein corresponding to human WASH complex subunit 7.
Purification Method Protein G Purified
Quantity 100 μg
Primary or Secondary Primary
Gene ID (Entrez) NP_056090.1
Target Species Human
Content And Storage 2-8°C for 12 months
Form Purified
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.