Learn More
WBP1, Rabbit, Monoclonal Antibody, Abnova™
Rabbit monoclonal antibody raised against a human WBP1 peptide using ARM Technology.
Supplier: Abnova Corporation H00023559K
Includes: Up to three rabbit IgG clones of 1mg each will be delivered to customer.
Description
The globular WW domain is composed of 38 to 40 semiconserved amino acids shared by proteins of diverse functions including structural, regulatory, and signaling proteins. The domain is involved in mediating protein-protein interactions through the binding of polyproline ligands. This gene encodes a WW domain binding protein, which binds to the WW domain of Yes kinase-associated protein by a conserved region: XPPXY motif. The function of this protein has not been determined. [provided by RefSeq
Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)Expression: Overexpression vector and transfection into 293H cell line
Order Info
- Customer may provide cell or tissue lysate for antibody screening.
- Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.
Specifications
WBP1 | |
Monoclonal | |
Unconjugated | |
In 1X PBS, pH 7.4 | |
WBP1 | |
WBP1 | |
A synthetic peptide of human WBP1 | |
RUO | |
23559 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG |
ELISA, Western Blot | |
1mg | |
WW domain binding protein 1 | |
WBP1 | |
MGC15305, WBP-1 | |
Rabbit | |
1 mg x up to 3 Abs | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.