Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
WDR37 Rabbit anti-Human, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA526258
Description
This antibody is predicted to react with mouse based on sequence homology.
WDR37 encodes a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes.
Specifications
| WDR37 | |
| Polyclonal | |
| WDR37 | |
| RP11-529L18.2, WD repeat-containing protein 37 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 22884 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Q9Y2I8 | |
| WDR37 | |
| KLH conjugated synthetic peptide between 426-455 amino acids from the C-terminal region of human WDR37 | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction