Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WD repeat domain 42A, Mouse, Clone: 1A12, Abnova™

Catalog No. 89001539 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-001-539 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-001-539 Supplier Abnova Corporation Supplier No. H00050717M01A
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against a full-length recombinant WDR42A.

This gene encodes a WD repeat-containing protein that interacts with the Cul4-Ddb1 E3 ligase macromolecular complex. Multiple alternatively spliced transcript variants have been found for this gene. [provided by RefSeq

Sequence: MSSKGSSTDGRTDLANGSLSSSPEEMSGAEEGRETSSGIEVEASDLSLSLTGDDGGPNRTSTESRGTETESSGEDKDSDSMEDTGHYSINDENRVHDRSEEEEEEEEEEEEEQPRRRVQRKRANRDQDSSDDERALEDWVSSETSALPRPRWQALPALRERELGSSARFVYEACGARVFVQRFRLQHGLEGHTGCVNTLHFNQRGTWLASGSDDLKVVVWDWVRRQPVLDFESGHKSNVFQVRQGSIIATERIRHELEKSELQHGLGADSELL

Specifications

Antigen WD repeat domain 42A
Applications ELISA, Western Blot
Classification Monoclonal
Clone 1A12
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against a full length recombinant WDR42A.
Formulation ascites with no preservative
Gene WDR42A
Gene Accession No. BC013107
Gene Alias DKFZp781G1096/FLJ35857/H326/MGC117276/MGC118891/MGC99640
Gene Symbols WDR42A
Host Species Mouse
Immunogen WDR42A (AAH13107, 1 a.a. ∼ 273 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Quantity 200 μL
Regulatory Status RUO
Whole Molecule Yes
Primary or Secondary Primary
Gene ID (Entrez) 50717
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Ascites
Isotype IgM κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.