Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WDR5 Rabbit anti-Human, Polyclonal, MilliporeSigma™

Catalog No. 07706MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
07-706-MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 07-706-MI Supplier MilliporeSigma Supplier No. 07706
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Detect WDR5 using this Anti-WDR5 Antibody validated for use in western blotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WDR5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation Purified rabbit polyclonal IgG in buffer containing 0.1M Tris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide with 30% glycerol.
Gene Symbols BIG-3; BIG3; CFAP89; SWD3; WDR5
Host Species Rabbit
Immunogen KLH-conjugated linear peptide corresponding to the N-terminus of human WDR5.
Purification Method Affinity Purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) NP_060058
Target Species Human
Content And Storage −20°C in undiluted aliquots for 12 months. Avoid repeated freeze/thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.