Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

West Nile Virus/Kunjin Mouse anti-Human, Clone: 3.67G, MilliporeSigma™

Catalog No. MAB8150MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8150MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8150MI Supplier MilliporeSigma Supplier No. MAB8150
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-West Nile Virus/Kunjin Antibody, Envelope, clone 3.67G detects level of West Nile Virus/Kunjin and has been published and validated for use in ELISA, immunofluoresence, western blot, immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen West Nile Virus/Kunjin
Applications ELISA, Immunofluorescence, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 3.67G
Conjugate Unconjugated
Formulation Purified immunoglobulin in 0.02Mphosphate buffer, 0.25M NaCl, pH 7.6 with 0.1% Sodium Azide.
Host Species Mouse
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.