Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Western Equine Encephalitis Mouse anti-Human, Clone: 2A2C.3, MilliporeSigma™

Catalog No. MAB8742MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB8742MI 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB8742MI Supplier MilliporeSigma Supplier No. MAB8742
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect Western Equine Encephalitis using this Anti-Western Equine Encephalitis Antibody, clone 2A2C.3 validated for use in ELISA, immunofluoresence, HI.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Western Equine Encephalitis
Applications ELISA, Immunofluorescence
Classification Monoclonal
Clone 2A2C.3
Conjugate Unconjugated
Formulation Liquid; no preservatives
Host Species Mouse
Immunogen McMillan strain
Quantity 100 μL
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C in undiluted aliquots for up to 12 months.
Form Ascites
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.