Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WFDC3, Rabbit, Monoclonal Antibody, Abnova™

Catalog No. 89209107 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg x up to 3 Abs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-209-107 1 mg x up to 3 Abs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-209-107 Supplier Abnova Corporation Supplier No. H00140686K

Includes

Up to three rabbit IgG clones of 1mg each will be delivered to customer.

Rabbit monoclonal antibody raised against a human WFDC3 peptide using ARM Technology.

This gene encodes a member of the WAP-type four-disulfide core (WFDC) domain family. The WFDC domain, or WAP signature motif, contains eight cysteines forming four disulfide bonds at the core of the protein, and functions as a protease inhibitor. The encoded protein contains four WFDC domains. Most WFDC genes are localized to chromosome 20q12-q13 in two clusters: centromeric and telomeric. This gene belongs to the telomeric cluster. Alternatively spliced transcript variants have been observed but their full-length nature has not been determined. [provided by RefSeq

Library Construction: Nonfusion antibody library from rabbit spleen (ARM Technology)
Expression: Overexpression vector and transfection into 293H cell line

Order Info

  1. Customer may provide cell or tissue lysate for antibody screening.
  2. Rabbit monoclonal antibody generated by ARM technology is amenable to antibody engineering including F(ab)2, IgG, scFv and different Fc and non-Fc conjugates per customer request.

Specifications

Antigen WFDC3
Applications ELISA, Western Blot
Classification Monoclonal
Concentration 1mg
Conjugate Unconjugated
Description WAP four-disulfide core domain 3
Formulation In 1X PBS, pH 7.4
Gene WFDC3
Gene Accession No. WFDC3
Gene Alias WAP14, dJ447F3.3
Gene Symbols WFDC3
Host Species Rabbit
Immunogen A synthetic peptide of human WFDC3
Quantity 1 mg x up to 3 Abs
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 140686
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.