Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WHSC1/NSD2 Mouse anti-Human, Clone: 29D1, MilliporeSigma™

Catalog No. MABE191MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MABE191MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MABE191MI Supplier MilliporeSigma Supplier No. MABE191
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Detect WHSC1/NSD2 using this Anti-WHSC1/NSD2 Antibody, clone 29D1 validated for use in Western Blotting, Chromatin Immunoprecipitation (ChIP), Immunoprecipitation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WHSC1/NSD2
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 29D1
Conjugate Unconjugated
Formulation Purified mouse monoclonal IgG2bκ in buffer containing 0.1MTris-Glycine (pH 7.4), 150mM NaCl with 0.05% sodium azide.
Gene Accession No. NP_001035889
Gene Symbols WHSC1, MMSET, NSD2, TRX5
Host Species Mouse
Immunogen GST-tagged recombinant protein corresponding to human WHSC1/NSD2.
Purification Method Protein G purified
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C for one year
Form Purified
Isotype IgG2b κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.