Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wilms' Tumor Mouse anti-Human, Mouse, Clone: 6F-H2, MilliporeSigma™

Catalog No. MAB4234MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB4234MI 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MAB4234MI Supplier MilliporeSigma Supplier No. MAB4234
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This Anti-Wilms′ Tumor Antibody, NT clone 6F-H2, Ascites Free is validated for use in Immunocytochemistry, Immunoprecipitation, Immunofluorescence, Immunohistochemistry (Paraffin), and Western Blotting for the detection of Wilms′ tumor protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Wilms' Tumor
Applications ELISA, Immunofluorescence, Western Blot
Classification Monoclonal
Clone 6F-H2
Conjugate Unconjugated
Formulation Purified immunoglobulin from Protein A Sepharose™ chromatography. Liquid in 0.02M PB, pH 7.6, 0.25M NaCl containing 0.1% sodium azide.
Gene Accession No. NM_000378.3; NM_024424.2; NM_024425.2; NM_024426.3
Gene Symbols WT1, GUD, WT33, WAGR, WIT-2
Host Species Mouse
Immunogen aa 1-173 amino acides of the WT-1 protein
Quantity 100 μg
Primary or Secondary Primary
Target Species Human
Content And Storage 2-8°C in undiluted aliquots for up to 6 months.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.