Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WIP (pS488) Mouse, PE, Clone: K32-824, BD

Catalog No. BDB558673 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB558673 50 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB558673 Supplier BD Biosciences Supplier No. 558673
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Wiskott-Adrich syndrome protein (WASP)-Interacting Protein (WIP) is a member of the verprolin family of proteins that regulate cytoskeletal organization in a wide variety of cellular activities, including endocytosis, cellular adhesion and migration, mast cell degranulation, and lymphocyte activation. The 503-amino acid WIP protein contains binding sites for actin (globular and filamentous) and other proteins that are involved in the regulation of actin polymerization, such as WASP, N-WASP, profilin, cortactin, Hck, and NCK. As its functions imply, WIP is localized in actin-rich cell structures.

Intracellular Staining

TRUSTED_SUSTAINABILITY

Specifications

Antigen WIP (pS488)
Applications Flow Cytometry
Classification Monoclonal
Clone K32-824
Conjugate PE
Description PRPL-2 protein, WAIP, WASIP, WASPIP
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Host Species Mouse
Immunogen Phosphorylated Human WIP Peptide
Purification Method Affinity Purified
Quantity 50 Tests
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.