Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

wingless-type MMTV integration site family, member 8A, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89016932 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-016-932 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-016-932 Supplier Abnova Corporation Supplier No. H00007478B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human WNT8A protein.

The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family, and may be implicated in development of early embryos as well as germ cell tumors. It encodes a protein which shows 81% amino acid identity to the mouse Wnt8A protein. [provided by RefSeq

Sequence: MGNLFMLWAALGICCAAFSASAWSVNNFLITGPKAYLTYTTSVALGAQSGIEECKFQFAWERWNCPENALQLSTHNRLRSATRETSFIHAISSAGVMYIITKNCSMGDFENCGCDGSNNGKTGGHGWIWGGCSDNVEFGERISKLFVDSLEKGKDARALMNLHNNRAGRLAVRATMKRTCKCHGISGSCSIQTCWLQLAEFREMGDYLKAKYDQALKIEMDKRQLRAGNSAEGHWVPAEAFLPSAEAELIFLEESPDYCTCNSSLGIYGTEGRECLQNSHNTSRWERRSCGRLCTECGLQVEERKTEVISSCNCKFQWCCTVKCDQCRHVVSKYYCARSPGSAQSLGKGSA

Specifications

Antigen wingless-type MMTV integration site family, member 8A
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human WNT8A protein.
Formulation PBS with no preservative; pH 7.4
Gene WNT8A
Gene Accession No. NM_058244
Gene Alias WNT8D
Gene Symbols WNT8A
Host Species Mouse
Immunogen WNT8A (NP_490645, 1 a.a. ∼ 351 a.a) full-length human protein.
Purification Method Affinity Purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 7478
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.