Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WRB, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002806 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-806 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-806 Supplier Abnova Corporation Supplier No. H00007485B02P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human WRB protein.

This gene encodes a basic nuclear protein of unknown function. The gene is widely expressed in adult and fetal tissues. Since the region proposed to contain the gene(s) for congenital heart disease (CHD) in Down syndrome (DS) patients has been restricted to 21q22.2-22.3, this gene, which maps to 21q22.3, has a potential role in the pathogenesis of Down syndrome congenital heart disease. Alternatively spliced transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq

Sequence: MSSAAADHWAWLLVLSFVFGCNVLRILLPSFSSFMSRVLQKDAEQESQMRAEIQDMKQELSTVNMMDEFARYARLERKINKMTDKLKTHVKARTAQLAKIKWVISVAFYVLQAALMISLIWKYYSVPVAVVPSKWITPLDRLVAFPTRVAGGVGITCWILVCNKVVAIVLHPFS

Specifications

Antigen WRB
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human WRB protein.
Formulation PBS with no preservative; pH 7.4
Gene WRB
Gene Accession No. NM_004627.2
Gene Alias CHD5
Gene Symbols WRB
Host Species Mouse
Immunogen WRB (NP_004618.2, 1 a.a. ∼ 174 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7485
Target Species Rat
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.