Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WT1 Mouse anti-Human, Clone: 6F-H2, MilliporeSigma™

Catalog No. 05753MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
05-753-MI 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 05-753-MI Supplier MilliporeSigma Supplier No. 05753
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Anti-WT1 Antibody, clone 6F-H2 is a Mouse Monoclonal Antibody for detection of WT1 also known as Wilms′ Tumor and has been tested in western blot, immunocytochemistry and immunohistochemistry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen WT1
Applications Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6F-H2
Conjugate Unconjugated
Formulation 0.1M Tris-glycine, pH 7.4, 0.15M NaCl, 0.05% sodium azide before the addition of glycerol to 30%
Gene Accession No. NM_024426
Gene Symbols WT1, WAGR, GUD, WT33, WIT-2
Host Species Mouse
Immunogen 6His-tagged fusion protein corresponding to residues 1-181 of human WT1 (Wilms tumor)
Purification Method Protein G purfied
Quantity 200 μg
Primary or Secondary Primary
Target Species Human
Content And Storage −20°C for 24 months.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.