Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

WWOX, Mouse, Purified MaxPab™ Polyclonal Antibody, Abnova™

Catalog No. 89002845 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-002-845 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-002-845 Supplier Abnova Corporation Supplier No. H00051741B01P
Add to Cart
Add to Cart

Mouse polyclonal antibody raised against a full-length human WWOX protein.

WW domain-containing proteins are found in all eukaryotes and play an important role in the regulation of a wide variety of cellular functions such as protein degradation, transcription, and RNA splicing. This gene encodes a protein which contains 2 WW domains and a short-chain dehydrogenase/reductase domain (SRD). The highest normal expression of this gene is detected in hormonally regulated tissues such as testis, ovary, and prostate. This expression pattern and the presence of an SRD domain suggest a role for this gene in steroid metabolism. The encoded protein is more than 90% identical to the mouse protein, which is an essential mediator of tumor necrosis factor-alpha-induced apoptosis, suggesting a similar, important role in apoptosis for the human protein. In addition, there is evidence that this gene behaves as a suppressor of tumor growth. Alternative splicing of this gene generates transcript variants that encode different isoforms. [provided by RefSeq

Sequence: MAALRYAGLDDTDSEDELPPGWEERTTKDGWVYYANHTEEKTQWEHPKTGKRKRVAGDLPYGWEQETDENGQVFFVDHINKRTTYLDPRLAFTVDDNPTKPTTRQRYDGSTTAMEILQGRDFTGKVVVVTGANSGIGFETAKSFALHGAHVILACRNMARASEAVSRILEEWKTKYHPPPEKCRIKIFH

Specifications

Antigen WWOX
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Mouse polyclonal antibody raised against a full-length human WWOX protein.
Formulation PBS with no preservative; pH 7.4
Gene WWOX
Gene Accession No. NM_130791.1
Gene Alias D16S432E/FOR/FRA16D/HHCMA56/PRO0128/SDR41C1/WOX1
Gene Symbols WWOX
Host Species Mouse
Immunogen WWOX (NP_570607.1, 1 a.a. ∼ 189 a.a) full-length human protein.
Purification Method Affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51741
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.