Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ XAB2 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA14266

Catalog No. PA14266


Only null left
Add to Cart

Description

Description

PA1-4266 detects the XAB2 (XPA-binding protein 2) protein in human samples. PA1-4266 has been successfully used in Western blot procedures. PA1-4266 immunizing peptide corresponds to amino acid residues 840-855 of the human XAB2 protein.

A novel protein designated XAB2 (XPA-binding protein 2) interacts with XPA, a factor central to both nucleotide excision repair subpathways. Data suggests that XAB2 is a novel component involved in transcription-coupled repair and transcription.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

XAB2
Polyclonal
Unconjugated
XAB2
0610041O14Rik; Adapter protein ATH-55; Ath55; AV025587; crn-related protein kim1; DKFZp762C1015; HCNP; HCNP protein; HCRN; KIAA1177; NTC90; PP3898; Pre-mRNA-splicing factor SYF1; Protein HCNP; SYF1; SYF1 homolog, RNA splicing factor; SYF1 pre-mRNA-splicing factor; Xab2; XPA binding protein 2; XPA-binding protein 2
Rabbit
Antigen Affinity Chromatography
RUO
56949
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9HCS7
XAB2
Synthetic peptide: E(840) Q Q S V P A A V F G S L K E D(855).
100 μg
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.